missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSPAN31 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563445

Product Code. 15731222

  • € 566.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DVINASWWVM SNKTRDELER SFDCCGLFNL TTLYQQDYDF CTAICKSQSP TCQMCGEKF Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 71%.

TSPAN31 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is thought to be involved in growth-related cellular processes. It is associated with tumorigenesis and osteosarcoma.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TSPAN31
Polyclonal
Unconjugated
TSPAN31
2700085A14Rik; AW742554; sarcoma amplified sequence; sarcoma-amplified sequence; Sarcoma-amplified sequence homolog; SAS; tetraspanin 31; tetraspanin-31; transmembrane 4 protein; transmembrane protein; TSPAN31; Tspan-31; Unknown (protein for MGC:133819)
Rabbit
Antigen affinity chromatography
RUO
6302
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q12999
TSPAN31
Recombinant protein fragment of Human TSPAN31 (Product #RP-95638)
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.