missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN32 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582869
This item is not returnable.
View return policy
Description
Immunogen sequence: LTPRACGRQP QEPSLLRCSQ GGPTHCLHSE AVAIGPRGCS GSLRWLQESD AAPLPLSCHL AAHRALQGRS RGGLSGCP Highest antigen sequence indentity to the following orthologs: Mouse - 28%, Rat - 29%.
This gene, which is a member of the tetraspanin superfamily, is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of chromosome 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian and breast cancers. This gene is located among several imprinted genes; however, this gene, as well as the tumor-suppressing subchromosomal transferable fragment 4, escapes imprinting. This gene may play a role in malignancies and diseases that involve this region, and it is also involved in hematopoietic cell function. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
| TSPAN32 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN32 | |
| AML1-regulated transmembrane protein 1; ART1; Art-1; AW208513; BB235973; D7Wsu37e; FLJ17158; FLJ97586; MGC22455; pan hematopoietic expression; pan-hematopoietic expression protein; Phemx; PHMX; protein Phemx; tetraspanin; tetraspanin 32; tetraspanin-32; TSPAN32; Tspan-32; Tssc6; tumor-suppressing STF cDNA 6; tumor-suppressing subchromosomal transferable fragment 6; tumor-suppressing subchromosomal transferable fragment cDNA 6; tumor-suppressing subtransferable candidate 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10077 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96QS1 | |
| TSPAN32 | |
| Recombinant protein corresponding to Human TSPAN32. Recombinant protein control fragment (Product #RP-90918). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction