missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN33 Monoclonal Antibody (2F8A12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548821
This item is not returnable.
View return policy
Description
TSPAN33 Monoclonal Antibody for Western Blot, Flow, ELISA
TSPAN33 plays an important role in normal erythropoiesis. It has a role in the differentiation of erythroid progenitors.
Specifications
| TSPAN33 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q86UF1 | |
| TSPAN33 | |
| Purified recombinant fragment of human TSPAN33 (AA: Extra(46-64)+Extra(118-235)) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Western Blot | |
| 2F8A12 | |
| Unconjugated | |
| TSPAN33 | |
| 1300010A20Rik; hPen; hypothetical protein LOC539284; mPen; PEN; PEN.; Penumbra; proerythroblast new membrane; RGD1560915; tetraspanin 33; tetraspanin-33; Tetraspanin-33-like protein; Tspan33; tspan-33 | |
| Mouse | |
| Protein G | |
| RUO | |
| 340348 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction