missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN6 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | TSPAN6 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TSPAN6 Polyclonal antibody specifically detects TSPAN6 in Mouse samples. It is validated for Western BlotSpecifications
| TSPAN6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 7105 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| Putative NF-kappa-B-activating protein 321, T245, T245 protein, tetraspanin 6, Tetraspanin TM4-D, tetraspanin-6, TM4SF6tetraspan TM4SF, Transmembrane 4 superfamily member 6A15 homolog, TSPAN-6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-200 of human TSPAN6 (NP_003261.1). NSFKNNYEKALKQYNSTGDYRSHAVDKIQNTLHCCGVTDYRDWTDTNYYSEKGFPKSCCKLEDCTPQRDADKVNNEGCFIK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title