missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN7 Monoclonal Antibody (OTI2D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811638S
This item is not returnable.
View return policy
Description
TSPAN7 Monoclonal antibody specifically detects TSPAN7 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| TSPAN7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P56177 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1745 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D6 | |
| Unconjugated | |
| DLX1 | |
| 1200014P11Rik; A15; AI323365; CCG-B7; CD231; CD231 antigen; cell surface glycoprotein A15; DXS1692E; membrane component chromosome X surface marker 1; membrane component, X chromosome, surface marker 1; MRX58; Mxs1; neuroprotective protein 5; PE31; R74651; TALLA; TALLA homolog; TALLA-1; T-cell acute lymphoblastic leukemia associated antigen 1; T-cell acute lymphoblastic leukemia-associated antigen 1; tetraspanin 7; tetraspanin protein; tetraspanin-7; TM4SF2; TM4SF2b; transmembrane 4 superfamily 2b; transmembrane 4 superfamily member 2; Transmembrane 4, superfamily member 2 (cell surface glycoprotein A15); transmembrane protein A15; TSPAN7; tspan-7 | |
| Human recombinant protein fragment corresponding to amino acids 1-255 of human DLX1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction