Learn More
Novus Biologicals™ TSPAN8/TM4SF3 Antibody (INSERM patent anti-CO-029) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28263-1mg
 View more versions of this product
Product Code. 30046126
  • € 920.00 / 1mg
Description
TSPAN8/TM4SF3 Monoclonal antibody specifically detects TSPAN8/TM4SF3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TSPAN8/TM4SF3
Monoclonal
Unconjugated
P19075
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
INSERM patent anti-CO-029
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CO-029, tetraspanin 8, TM4SF3tspan-8, Transmembrane 4 superfamily member 3tetraspanin-8, Tspan-8, Tumor-associated antigen CO-029
TSPAN8
1 mg
Cancer
7103