missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPO/PBR Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | TSPO/PBR |
|---|---|
| Clone | 1D1F12 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236079
|
Proteintech
68137-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233194
|
Proteintech
68137-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TSPO/PBR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TSPO/PBR. This antibody recognizes human, mouse, rat antigen. The TSPO/PBR antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| TSPO/PBR | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 706 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1D1F12 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| BZRP; DBI; IBP; MBR; PBR; PKBS; PTBR; mDRC; pk18 | |
| IgM | |
| Caprylic acid/ammonium sulfate precipitation | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title