missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPY1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548721
This item is not returnable.
View return policy
Description
TSPY1 Polyclonal Antibody for Western Blot, IHC (P)
May be involved in sperm differentiation and proliferation.
Specifications
| TSPY1 | |
| Polyclonal | |
| Unconjugated | |
| TSPY1 | |
| Cancer/testis antigen 78; CT78; DYS14; pJA923; testis specific protein, Y-linked 1; testis-specific Y-encoded protein 1; TSPY; TSPY1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 7258 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q01534 | |
| TSPY1 | |
| KLH conjugated synthetic peptide between 274-303 amino acids from the C-terminal region of human TSPY. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction