missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSPY3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA548840

Product Code. 15663135

  • € 520.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TSPY3 Polyclonal Antibody for Western Blot

TSPY3 (Testis Specific Protein Y-Linked 3) is a Protein Coding gene. An important paralog of this gene is TSPY4. [GeneCards].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TSPY3
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
P0CV98
TSPY3
KLH conjugated synthetic peptide between 277-305 amino acids from the C-terminal region of human TSPY3.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
TSPY3
Cancer/testis antigen 78; CT78; testis specific protein, Y-linked 3; Testis-specific Y-encoded protein 1; Testis-specific Y-encoded protein 10; Testis-specific Y-encoded protein 3; TSPY; TSPY1; TSPY10; TSPY3
Rabbit
Antigen affinity chromatography, Protein A
RUO
728137
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.