missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPY3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22257-1-AP-20UL
This item is not returnable.
View return policy
Description
Unknown. May be involved in sperm differentiation and proliferation.Specifications
| TSPY3 | |
| Polyclonal | |
| Unconjugated | |
| TSPY3 | |
| Cancer/testis antigen 78, CT78, TSPY, TSPY1, TSPY3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 728137 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.56 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P0CV98 | |
| TSPY3 | |
| TSPY3 Fusion Protein Ag17762 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction