missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSR1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15918504
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15918504 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15918504 Supplier Invitrogen™ Supplier No. PA565764

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TILFLLDPLEG WDSTGDYCLS CLFAQGLPTY TLAVQGISGL PLKKQIDTRK KLSKAVEKRF PHDKLLLLDT QQEAGMLLRQ LANQKQQHLA FRDRRAYLFA HAVDFVPSEE NNLVGTLKIS GYVRGQTLNV NRLLHIVGYG DFQM Highest antigen sequence identity to the following orthologs - mouse 90%, rat 85%.

Required during maturation of the 40S ribosomal subunit in the nucleolus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSR1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSR1
Gene Accession No. Q2NL82
Gene Alias AU040765; AW550801; KIAA1401; mKIAA1401; pre-rRNA-processing protein TSR1 homolog; Tsr1; TSR1 20S rRNA accumulation; TSR1, 20S rRNA accumulation, homolog; TSR1, 20S rRNA accumulation, homolog (S. cerevisiae); TSR1, ribosome maturation factor
Gene Symbols TSR1
Host Species Rabbit
Immunogen Recombinant Human TSR1. Recombinant protein control fragment (Product #RP-104962).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55720
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.