missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583514
This item is not returnable.
View return policy
Description
Immunogen sequence: FLANAKESPQ EEEIDPFDVD SGREFGNPNR PVASTRLPSD TDDSDASEDP Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 68%.
TSR3 is a probable pre-rRNA processing protein involved in ribosome biogenesis.
Specifications
| TSR3 | |
| Polyclonal | |
| Unconjugated | |
| TSR3 | |
| C16orf42; MGC24381; probable ribosome biogenesis protein C16orf42; ribosome biogenesis protein TSR3 homolog; TSR3; TSR3, 20S rRNA accumulation, homolog; TSR3, acp transferase ribosome maturation factor; UND313L | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115939 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UJK0 | |
| TSR3 | |
| Recombinant protein corresponding to Human TSR3. Recombinant protein control fragment (Product #RP-98611). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction