missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSSK4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 624.00
Specifications
| Antigen | TSSK4 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18244802
|
Novus Biologicals
NBP2-56284 |
100 μL |
€ 624.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628847
|
Novus Biologicals
NBP2-56284-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TSSK4 Polyclonal specifically detects TSSK4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| TSSK4 | |
| Polyclonal | |
| Rabbit | |
| Protein Phosphatase | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 283629 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PFDDTNLKKLLRETQKEVTFPANHTISQECKNLILQMLRQATKRATILDIIKDSWVLKFQPEQPTHEIRLLEAMCQLH | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| C14orf20, EC 2.7.11, EC 2.7.11.1, MGC133264, serine/threonine kinase 22E, Serine/threonine-protein kinase 22E, STK22E, Testis-specific kinase 4, testis-specific serine kinase 4, testis-specific serine/threonine-protein kinase 4, TSK-4, TSSK-4, TSSK5TSK4 | |
| TSSK4 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title