missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSSK6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567708
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. TSSK 6 (testis-specific serine kinase 6), also known as SSTK, TSSK4, FKSG82 or CT72, is a 273 amino acid protein that contains one protein kinase domain and belongs to the Ser/Thr protein kinase family. Highly expressed in testis with lower expression in ovary, colon, thymus, small intestine and spleen, TSSK 6 catalyzes the ATP-dependent phosphorylation of proteins involved in sperm production and chromatin remodeling. TSSK 6 uses magnesium as a cofactor and is thought to be required for proper sperm development and function, as well as DNA condensation events. Defects in the gene encoding TSSK 6 are associated with male infertility characterized by low sperm count and decreased sperm motility.
Specifications
| TSSK6 | |
| Polyclonal | |
| Unconjugated | |
| TSSK6 | |
| Cancer/testis antigen 72; CT72; FKSG82; RGD1559764; serine/threonine protein kinase SSTK; serine/threonine-protein kinase SSTK; Small serine/threonine kinase; SSTK; testis secretory sperm-binding protein Li 205a; testis specific serine kinase 6; testis-specific kinase 6; testis-specific serine kinase 6; testis-specific serine/threonine-protein kinase 6; TSK-6; TSSK4; TSSK6; TSSK-6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 290670, 83983, 83984 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q925K9, Q9BXA6 | |
| TSSK6 | |
| Synthesized peptide derived from human TSSK6. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction