Learn More
TST Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-20752
Product Code. 18146711
  • ONLINE EXCLUSIVE PRICE
    € 629.00
    € 594.30 / 0.10mL
    Save € 34.70
    5% Off
Description
TST Polyclonal specifically detects TST in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TST
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
EC 2.8.1, EC 2.8.1.1, MGC19578, RDSRhodanese, thiosulfate sulfurtransferase, thiosulfate sulfurtransferase (rhodanese)
Rabbit
33 kDa
0.1 mL
Proteases & Other Enzymes
7263
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
0.71mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Q16762
TST
Recombinant protein encompassing a sequence within the center region of human TST. The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Human, Mouse, Rat