missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSTA3 Polyclonal Antibody
GREENER_CHOICE

Product Code. p-200129683
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17780864 200 μL 200µL
17772914 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17780864 Supplier Invitrogen™ Supplier No. PA5119464

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TSTA3 Polyclonal Antibody for Western Blot, IHC (P)

Tissue specific transplantation antigen P35B is a NADP(H)-binding protein. It catalyze the two-step epimerase and the reductase reactions in GDP-D-mannose metabolism, converting GDP-4-keto-6-D-deoxymannose to GDP-L-fucose. GDP-L-fucose is the substrate of several fucosyltransferases involved in the expression of many glycoconjugates, including blood group ABH antigens and developmental adhesion antigens. Mutations in this gene may cause leukocyte adhesion deficiency, type II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSTA3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TSTA3
Gene Accession No. P23591, Q13630
Gene Alias 3-5 epimerase/4-reductase; AI256181; FX; GDP-4-keto-6-deoxy-D-mannose epimerase-reductase; GDP-4-keto-6-deoxy-D-mannose-3,5-epimerase-4-reductase; GDP-fucose synthase; GDP-L-fucose synthase; GDP-L-fucose synthetase; GFUS; LOC513158 protein; P35B; Protein FX; red cell NADP(H)-binding protein; SDR4E1; short chain dehydrogenase/reductase family 4E, member 1; Short-chain dehydrogenase/reductase family 4E member 1; testis tissue sperm-binding protein Li 45a; tissue specific transplantation antigen 3; tissue specific transplantation antigen P35B; transplantation antigen P35B; Tsta3; Tstap35b; Tum-P35B antigen
Gene Symbols TSTA3
Host Species Rabbit
Immunogen Recombinant protein Tissue Specific Transplantation Antigen P35B. The antigen corresponds to amino acid range 1-321 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22122, 300036, 7264
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.