missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSTD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552316
This item is not returnable.
View return policy
Description
Immunogen sequence: MAGAPTVSLP ELRSLLASGR ARLFDVRSRE EAAAGTIPGA LNIPVSELES ALQMEPAAFQ ALYSAEKPKL EDEHLVFFCQ MGKRGLQATQ LARSLGYTGY G Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.
Thiosulfate:glutathione sulfurtransferase (TST) required to produce S-sulfanylglutathione (GSS(-)), a central intermediate in hydrogen sulfide metabolism (PubMed:24981631). Provides the link between the first step in mammalian H(2)S metabolism performed by the sulfide:quinone oxidoreductase (SQOR) which catalyzes the conversion of H(2)S to thiosulfate, and the sulfur dioxygenase (SDO) which uses GSS(-) as substrate (PubMed:24981631). The thermodynamic coupling of the irreversible SDO and reversible TST reactions provides a model for the physiologically relevant reaction with thiosulfate as the sulfane donor (PubMed:24981631). [UniProt].
Specifications
| TSTD1 | |
| Polyclonal | |
| Unconjugated | |
| TSTD1 | |
| KAT; putative thiosulfate sulfurtransferase KAT; thiosulfate sulfurtransferase (rhodanese)-like domain containing 1; thiosulfate sulfurtransferase KAT, putative; thiosulfate sulfurtransferase like domain containing 1; thiosulfate sulfurtransferase/rhodanese-like domain-containing protein 1; Thiosulfate:glutathione sulfurtransferase; TST; TSTD1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100131187 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFU3 | |
| TSTD1 | |
| Recombinant protein corresponding to Human TSTD1. Recombinant protein control fragment (Product #RP-89177). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction