missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTBK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 572.00
Specifications
| Antigen | TTBK1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18228041
|
Novus Biologicals
NBP2-55410 |
100 μL |
€ 572.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619486
|
Novus Biologicals
NBP2-55410-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TTBK1 Polyclonal specifically detects TTBK1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| TTBK1 | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 84630 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LPPQLSQGDGRSETSQPPTPGSPSHSPLHSGPRPRRRESDPTGPQRQVFSVAPPFEVNGLPRAVPLSLPYQDFKRDLSDYRERARLLNRVR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| BDTK, Brain-derived tau kinase, EC 2.7.11, EC 2.7.11.1, KIAA1855tau-tubulin kinase 1, RP3-330M21.4, tau tubulin kinase 1 | |
| TTBK1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title