missing translation for 'onlineSavingsMsg'
Learn More

TTC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16878913 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16878913 20 μL 20µL
16868913 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16878913 Supplier Proteintech Supplier No. 116761AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a protein that belongs to the tetratrico peptide repeat superfamily of proteins. The encoded protein plays a role in protein-protein interactions, and binds to the Galpha subunit of G protein-coupled receptors to activate the Ras signaling pathway. Alternate splicing results in multiple transcript variants.

Specifications

Antigen TTC1
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TTC1
Gene Accession No. Q91Z38, Q99614
Gene Alias TPR repeat protein 1, TPR1, TTC1
Gene Symbols Ttc1
Host Species Rabbit
Immunogen TTC1 Fusion Protein Ag2277
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287208, 66827, 7265
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.