missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97168-100ul
This item is not returnable.
View return policy
Description
TTC11 Polyclonal antibody specifically detects TTC11 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| TTC11 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:50-1:200 | |
| CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11 | |
| Produced in rabbits immunized with E. coli-derived Human TTC11 fragment. | |
| 100 μg | |
| Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors | |
| 51024 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction