missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 724.00
Specifications
| Antigen | TTC11 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228091
|
Novus Biologicals
NBP3-37945-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227562
|
Novus Biologicals
NBP3-37945-100ul |
100 μL |
€ 724.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TTC11 Polyclonal antibody specifically detects TTC11 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| TTC11 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors | |
| PBS (pH 7.3), 50% glycerol | |
| 51024 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-122 of human TTC11 (NP_057152.2).,, Sequence:, MEAVLNELVSVEDLLKFEKKFQSEKAAGSVSKSTQFEYAWCLVRSKYNDDIRKGIVLLEELLPKGSKEEQRDYVFYLAVGNYRLKEYEKALKYVRGLLQTEPQNNQAKELERLIDKAMKKDG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title