missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC11 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97168PCP
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
TTC11 Polyclonal antibody specifically detects TTC11 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Tekniske data
| TTC11 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry-Paraffin | |
| CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11 | |
| Produced in rabbits immunized with E. coli-derived Human TTC11 fragment. | |
| 0.1 mL | |
| Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors | |
| 51024 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion