missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC14 Monoclonal Antibody (OTI1G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808787S
This item is not returnable.
View return policy
Description
TTC14 Monoclonal antibody specifically detects TTC14 in Human samples. It is validated for Western BlotSpecifications
| TTC14 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P20794 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4117 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1G9 | |
| Unconjugated | |
| MAK | |
| 2700016E08Rik; 4930434D01Rik; 4931403I22Rik; 4933402I15Rik; AI662468; AU014779; AW561908; cI-44; DRDL5813; Kiaa1980; mKIAA1980; PRO19630; tetratricopeptide repeat domain 14; tetratricopeptide repeat protein 14; TPR repeat protein 14; Ttc14; UNQ5813; UNQ5813/PRO19630 | |
| Human recombinant protein fragment corresponding to amino acids 291-623 of human MAK produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction