missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC19 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20875-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein of unknown function. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined.Specifications
| TTC19 | |
| Polyclonal | |
| Unconjugated | |
| TTC19 | |
| 2010204O13Rik, TPR repeat protein 19, TTC19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54902, 72795 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6DKK2, Q8CC21 | |
| TTC19 | |
| TTC19 Fusion Protein Ag14974 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction