missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-84292
This item is not returnable.
View return policy
Description
TTC3 Polyclonal specifically detects TTC3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TTC3 | |
| Polyclonal | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| DCRR1tetratricopeptide repeat protein 3 (TPR repeat protein D)10RNF105DKFZp686M0150, E3 ubiquitin-protein ligase TTC3, EC 6.3.2.-, Protein DCRR1, RING finger protein 105, tetratricopeptide repeat domain 3, Tetratricopeptide repeat protein 3, TPR repeat protein 3, TPR repeat protein D, TPRDI, TPRDII, TPRDTPRDIII | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TTC3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DCPHVDDCVFAAEFMSNDYVRVTQLYCDGVGVQYKDYIQSERNLEFDICSIWCSKPISVLQDYCDAIKINIFWPLLFQHQNSSVISRLHPCVDANNSRASEINLKKLQHLELMEDIVDLAKKVANDS | |
| 0.1 mL | |
| Zinc Finger | |
| 7267 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction