Learn More
TTC32 Monoclonal Antibody (OTI5C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501341S
Encompass_Preferred
 View more versions of this product
Product Code. 17503971
  • € 242.00 / 30µL
Description
TTC32 Monoclonal antibody specifically detects TTC32 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
TTC32
Monoclonal
0.55 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q5I0X7
Mouse
Affinity chromatography
RUO
130502
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI5C12
Unconjugated
Ttc32
1700013G20Rik; tetratricopeptide repeat domain 32; tetratricopeptide repeat protein 32; TPR repeat protein 32; TTC32
Full length human recombinant protein of human TTC32 produced in HEK293T cell.