missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC32 Mouse anti-Human, Clone: OTI3C2, lyophilized, TrueMAB™
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TTC32 is a protein coding gene.
Specifications
Specifications
| Antigen | TTC32 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3C2 |
| Conjugate | Unconjugated |
| Formulation | PBS with 8% trehalose and no preservative; pH 7.3 |
| Gene | TTC32 |
| Gene Accession No. | Q5I0X7 |
| Gene Alias | tetratricopeptide repeat domain 32 |
| Gene Symbols | TTC32 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?