missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC39B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49150
This item is not returnable.
View return policy
Description
TTC39B Polyclonal antibody specifically detects TTC39B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TTC39B | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| C9orf52, chromosome 9 open reading frame 52, FLJ33868, tetratricopeptide repeat domain 39B, tetratricopeptide repeat protein 39B, TPR repeat protein 39B | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LKGCCLKNLQRPLQAELCYNHVVESEKLLKYDHYLVPFTLFELASLYKSQGEIDKAIKFLETARNNYKDYS | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 158219 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction