missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTC39B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563021
This item is not returnable.
View return policy
Description
Immunogen sequence: LKGCCLKNLQ RPLQAELCYN HVVESEKLLK YDHYLVPFTL FELASLYKSQ GEIDKAIKFL ETARNNYKDY S Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
Regulates high density lipoprotein (HDL) cholesterol metabolism by promoting the ubiquitination and degradation of the oxysterols receptors LXR (NR1H2 and NR1H3). [UniProt].
Specifications
| TTC39B | |
| Polyclonal | |
| Unconjugated | |
| TTC39B | |
| 1810054D07Rik; 9130422G05Rik; AI450603; AV103077; AW215585; C9orf52; T39; tetratricopeptide repeat domain 39B; tetratricopeptide repeat protein 39B; TPR repeat protein 39B; Ttc39b | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 158219 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VTQ0 | |
| TTC39B | |
| Recombinant protein corresponding to Human TTC39B. Recombinant protein control fragment (Product #RP-101376). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction