missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TTC39B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA566561

Product Code. 15989254

  • € 580.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SSSSTKVDLKS GLEECAVALN LFLSNKFTDA LELLRPWAKE SMYHALGYST IVVLQAVLTF EQQDIQNGIS AMKDALQ Highest antigen sequence identity to the following orthologs - mouse 92%, rat 91%.

Regulates high density lipoprotein (HDL) cholesterol metabolism by promoting the ubiquitination and degradation of the oxysterols receptors LXR (NR1H2 and NR1H3). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TTC39B
Polyclonal
Unconjugated
TTC39B
1810054D07Rik; 9130422G05Rik; AI450603; AV103077; AW215585; C9orf52; T39; tetratricopeptide repeat domain 39B; tetratricopeptide repeat protein 39B; TPR repeat protein 39B; Ttc39b
Rabbit
Antigen Affinity Chromatography
RUO
158219
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5VTQ0
TTC39B
Recombinant Human TTC39B. Recombinant protein control fragment (Product #RP-107216).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.