missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC4 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18426R
This item is not returnable.
View return policy
Description
TTC4 Polyclonal antibody specifically detects TTC4 in Human samples. It is validated for Western Blot
Specifications
| TTC4 | |
| Polyclonal | |
| Western Blot | |
| DKFZp781B0622, FLJ41930, MGC5097, tetratricopeptide repeat domain 4, tetratricopeptide repeat protein 4, TPR repeat protein 4 | |
| Recognizes region between residues Between Between 1 and 50.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7268 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction