missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11878-1-AP-150UL
This item is not returnable.
View return policy
Description
Effector protein for Rho-type GTPases. Regulates actin filament reorganization via its interaction with the Arp2/3 complex. Important for efficient actin polymerization. Possible regulator of lymphocyte and platelet function. Mediates actin filament reorganization and the formation of actin pedestals upon infection by pathogenic bacteria.Specifications
| TTC4 | |
| Polyclonal | |
| Unconjugated | |
| TTC4 | |
| TPR repeat protein 4, TTC4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362556, 72354, 7268 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95801, Q8R3H9 | |
| Ttc4 | |
| TTC4 Fusion Protein Ag2474 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction