missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 178.50
Specifications
| Antigen | TTC5 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
TTC5 Polyclonal specifically detects TTC5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| TTC5 | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8N0Z6 | |
| 91875 | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human TTC5. The immunogen is located within the last 50 amino acids of TTC5. Amino Acid Squence: SSSQASATVASRPQ | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Strap, tetratricopeptide repeat domain 5, tetratricopeptide repeat protein 5, TPR repeat protein 5 | |
| TTC5 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title