Learn More
TTF-1/ NKX2-1 Antibody (8G7G3/1 + NX2.1/690), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-47773JF549
Product Code. 18637759
  • € 588.00 / 0.10mL
Description
TTF-1 / NKX2-1 Monoclonal antibody specifically detects TTF-1 / NKX2-1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TTF-1 / NKX2-1
Monoclonal
Janelia Fluor 549
benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2
Rat full length TTF-1/ NKX2-1 recombinant protein (8G7G3/1); Recombinant TTF-1/ NKX2-1 protein (NX2.1/690) (Uniprot: P43699)
0.1 mL
Membrane Trafficking and Chaperones, Tumor Biomarkers
7080
Store at 4°C in the dark.
IgG1 κ
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
8G7G3/1 + NX2.1/690
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Rat