missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTF-1/ NKX2-1 Antibody (NX2.1/690), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47772G
This item is not returnable.
View return policy
Description
TTF-1 / NKX2-1 Monoclonal antibody specifically detects TTF-1 / NKX2-1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| TTF-1 / NKX2-1 | |
| Monoclonal | |
| DyLight 488 | |
| benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2 | |
| Recombinant TTF-1/ NKX2-1 protein (Uniprot: P43699) | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones, Tumor Biomarkers | |
| 7080 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| NX2.1/690 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction