missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584221
This item is not returnable.
View return policy
Description
Immunogen sequence: VKRARVSGDD FSVPSKNSES TLFDSVEGDG AMMEEGVKSR PRQKKTQACL ASKHVQEAPR LEPANEEHNV ETAEDSEIRY LSADSGDADD SDADLGSAV Highest antigen sequence indentity to the following orthologs: Mouse - 44%, Rat - 44%.
TTF1 is a transcription termination factor that is localized to the nucleolus and plays a critical role in ribosomal gene transcription. The protein mediates the termination of RNA polymerase I transcription by binding to Sal box terminator elements downstream of pre-rRNA coding regions.
Specifications
| TTF1 | |
| Polyclonal | |
| Unconjugated | |
| Ttf1 | |
| AV245725; BCH; BHC; Homeobox protein NK-2 homolog A; homeobox protein Nkx-2.1; NK-2; NK2 homeobox 1; NK-2 homolog A; NKX2; NKX2.1; NKX2-1; NKX2A; NMTC1; RGD1565673; RNA polymerase I termination factor; RNA polymerase I transcription termination factor; T/EBP; TEBP; Thyroid nuclear factor 1; thyroid transcription factor 1; thyroid transcription factor 1-like; thyroid-specific enhancer-binding protein; TITF1; Transcription termination factor 1; transcription termination factor I; transcription termination factor, RNA polymerase I; TTF1; TTF-1; TTF-I | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15361 | |
| Ttf1 | |
| Recombinant protein corresponding to Human TTF1. Recombinant protein control fragment (Product #RP-103201). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction