missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTF2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97769B
This item is not returnable.
View return policy
Description
TTF2 Polyclonal specifically detects TTF2 in Human samples. It is validated for Western Blot.
Specifications
| TTF2 | |
| Polyclonal | |
| Western Blot | |
| EC 3.6.1, EC 3.6.4.-, F2, HuF2lodestar protein, human factor 2, Lodestar homolog, RNA polymerase II termination factor, Transcription release factor 2, transcription termination factor 2, transcription termination factor, RNA polymerase II | |
| Produced in rabbits immunized with E. coli-derived Human TTF2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 8458 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction