missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTI1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79035
This item is not returnable.
View return policy
Description
TTI1 Polyclonal specifically detects TTI1 in Human samples. It is validated for Immunoprecipitation.
Specifications
| TTI1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| KIAA0406SMG10, Protein SMG10, Tel two interacting protein 1, Tel2 interacting protein 1 homolog (S. pombe), TEL2-interacting protein 1 homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9675 | |
| Store at 4C. Do not freeze. |
| Immunoprecipitation | |
| 1.0 mg/mL | |
| Immunoprecipitation 2-10ug/mg | |
| NP_055472.1 | |
| TTI1 | |
| The immunogen recognized by this antibody maps to a region between residue 1039 and 1089 of human Tel2 Interacting Protein 1 using the numbering given in entry NP_055472.1 (GeneID 9675). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction