missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TTI1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15604785
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15604785 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15604785 Supplier Invitrogen™ Supplier No. PA550852

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total TTI1 protein.

Regulator of the DNA damage response (DDR). Part of the TTT complex that is required to stabilize protein levels of the phosphatidylinositol 3-kinase-related protein kinase (PIKK) family proteins. The TTT complex is involved in the cellular resistance to DNA damage stresses, like ionizing radiation (IR), ultraviolet (UV) and mitomycin C (MMC). Together with the TTT complex and HSP90 may participate in the proper folding of newly synthesized PIKKs. Promotes assembly, stabilizes and maintains the activity of mTORC1 and mTORC2 complexes, which regulate cell growth and survival in response to nutrient and hormonal signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTI1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TTI1
Gene Accession No. O43156, Q91V83
Gene Alias 2610036D13Rik; AI449463; KIAA0406; Protein SMG10; SMG10; smg-10; smg-10 homolog, nonsense mediated mRNA decay factor; Tel two interacting protein 1; Tel2 interacting protein 1 homolog; TEL2-interacting protein 1 homolog; TELO2 interacting protein 1; TELO2-interacting protein 1 homolog; TTI1
Gene Symbols TTI1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the N terminal of human TELO2 interacting protein 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75425, 9675
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.