missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTL Antibody (OTI3G5), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74668B
This item is not returnable.
View return policy
Description
TTL Monoclonal specifically detects TTL in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| TTL | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| TTL | |
| Full length recombinant protein of human TTL (NP_714923) produced in HEK293T cells. | |
| 0.1 mL | |
| 150465.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI3G5 | |
| Western Blot | |
| EC 6.3.2.25,2410003M22Rik, MGC46235, tubulin tyrosine ligase, tubulin-tyrosine ligase, tubulin--tyrosine ligase | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction