missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTL Monoclonal Antibody (OTI3G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502027
This item is not returnable.
View return policy
Description
Catalyzes the post-translational addition of a tyrosine to the C-terminal end of detyrosinated alpha-tubulin.Specifications
| TTL | |
| Monoclonal | |
| 0.42 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NG68 | |
| TTL | |
| Full length human recombit protein of human TTL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3G5 | |
| Unconjugated | |
| TTL | |
| tubulin tyrosine ligase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 150465 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction