missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119700
This item is not returnable.
View return policy
Description
Positive Control: Human heart tissue lysates, rat brain tissue, mouse brain tissue, SH-SY5Y. Subcellular Location: Nucleoplasm, Vesicles.
Catalyzes the post-translational addition of a tyrosine to the C-terminal end of detyrosinated alpha-tubulin.
Specifications
| TTL | |
| Polyclonal | |
| Unconjugated | |
| TTL | |
| 2410003M22Rik; 2700049H19Rik; AI848570; Ttl; tubulin tyrosine ligase; tubulin-tyrosine ligase; tubulin--tyrosine ligase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 150465, 171572, 69737 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P38585, Q8NG68, Q9QXJ0 | |
| TTL | |
| Synthetic peptide within human TTL aa 230-270. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction