Learn More
Invitrogen™ TTL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587576
Description
Immunogen sequence: MYTFVVRDEN SSVYAEVSRL LLATGHWKRL RRDNPRFNLM LGERNRLPFG RLGHEPGLVQ LVNYYRGADK LCRKASLVKL IKTSPELAES CTWFPESYVI YPTNLKTPVA PAQNGIQPPI SNSRTDEREF FLASYNRKKE DGEGNVWIAK SSAGAKGEGI LISSEASELL DFIDNQGQVH VIQKYLEHPL LLEPGHRKFD IRSWVLVDHQ YNIYLYREGV LRTASEPYHV DNFQDKTCHL TNHCIQKEYS KNYGKYEEGN EMFFKEFNQY; Positive Samples: SH-SY5Y, HeLa, SKOV3, Mouse brain, Mouse lung, Mouse heart, Rat kidney.
Catalyzes the post-translational addition of a tyrosine to the C-terminal end of detyrosinated alpha-tubulin.
Specifications
| TTL | |
| Polyclonal | |
| Unconjugated | |
| TTL | |
| 2410003M22Rik; 2700049H19Rik; AI848570; Ttl; tubulin tyrosine ligase; tubulin-tyrosine ligase; tubulin--tyrosine ligase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 150465, 171572, 69737 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.44 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P38585, Q8NG68, Q9QXJ0 | |
| TTL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human TTL (NP_7149231). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.