missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTLL12 Antibody (OTI7F11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74676H
This item is not returnable.
View return policy
Description
TTLL12 Monoclonal specifically detects TTLL12 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TTLL12 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TTLL12 | |
| Full length recombinant protein of human TTLL12 (NP_055955) produced in HEK293T cells. | |
| 0.1 mL | |
| 23170.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI7F11 | |
| Western Blot, Immunohistochemistry | |
| dJ526I14.2, KIAA0153FLJ41795, tubulin tyrosine ligase-like family, member 12, tubulin--tyrosine ligase-like protein 12 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction