missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTLL12 Monoclonal Antibody (OTI7F11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525022
This item is not returnable.
View return policy
Description
TTLL12 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
TTLL12 (tubulin tyrosine ligase-like family, member 12) is a 644 amino acid protein that contains one TTL domain. Of the fourteen members of the TTLL family that modify tubulin, TTLL12 is the least characterized member. TTLL12 is highly expressed in a multitude of metastatic prostate cancer cell lines, therefore, it is considered a target for tumor therapy. Overexpression of TTLL12 is suggested to alter chromosome ploidy, whereas downregulation of TTLL12 influence several post-translational modifications of tubulin.
Specifications
| TTLL12 | |
| Monoclonal | |
| 0.59 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14166 | |
| TTLL12 | |
| Full length human recombinant protein of TTLL12 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Rat, Canine, Monkey | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI7F11 | |
| Unconjugated | |
| TTLL12 | |
| BC055368; D430005B17; dJ526I14.2; Inactive tubulin--tyrosine ligase-like protein 12; KIAA0153; RGD1305319; TTL domain protein; ttll12; tubulin tyrosine ligase like 12; tubulin tyrosine ligase-like family member 12; tubulin tyrosine ligase-like family, member 12; tubulin--tyrosine ligase-like protein 12 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23170, 300105, 474474 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction