missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTLL12 Mouse anti-Canine, Human, Rat, Clone: OTI7F11, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500799
This item is not returnable.
View return policy
Description
TTLL12 (tubulin tyrosine ligase-like family, member 12) is a 644 amino acid protein that contains one TTL domain. Of the fourteen members of the TTLL family that modify tubulin, TTLL12 is the least characterized member. TTLL12 is highly expressed in a multitude of metastatic prostate cancer cell lines, therefore, it is considered a target for tumor therapy. Overexpression of TTLL12 is suggested to alter chromosome ploidy, whereas downregulation of TTLL12 influence several post-translational modifications of tubulin.Specifications
| TTLL12 | |
| Monoclonal | |
| 0.59 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14166 | |
| TTLL12 | |
| Full length human recombit protein of human TTLL12 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI7F11 | |
| Unconjugated | |
| TTLL12 | |
| dJ526I14.2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23170, 300105, 474474 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction