missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTLL4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142542
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Glutamylase which preferentially modifies beta-tubulin and non-tubulin proteins, such as NAP1L1, NAP1L4 and CGAS. Involved in the side-chain initiation step of the polyglutamylation reaction rather than in the elongation step. Involved in formation of short side-chains. Mediates initiation of polyglutamylation of nucleosome assembly proteins NAP1L1 and NAP1L4. Also acts as a monoglutamylase: generates monoglutamylation of CGAS, leading to impair the nucleotidyltransferase activity of CGAS. [UniProt].
Specifications
| TTLL4 | |
| Polyclonal | |
| Unconjugated | |
| TTLL4 | |
| KIAA0173; TTLL4; Tubulin polyglutamylase TTLL4; tubulin tyrosine ligase like 4; tubulin tyrosine ligase-like family member 4; tubulin tyrosine ligase-like family, member 4; tubulin--tyrosine ligase-like protein 4 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 488525 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| 0 | |
| TTLL4 | |
| Peptide with sequence C-RSSYRQKPYQQLE (aa 120-132). | |
| 100 μg | |
| Primary | |
| Canine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction