Learn More
TTYH3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24707-1-AP-20UL
 View more versions of this product
Product Code. 18079345
  • € 145.00 / 20µL
Description
The TTYH3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TTYH3. This antibody recognizes human antigen. The TTYH3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TTYH3
650 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
hTTY3, KIAA1691, Protein tweety homolog 3, TTYH3, tweety homolog 3 (Drosophila)
20 μL
80727
Store at -20°C. Stable for one year after shipment.