Learn More
Techne™ Advanced High-Temperature Digital Immersion Circulator
Product Code. 15375253
Click to view available options
Electrical Requirements:
120 V
Unit Size:
Each
Product Code. 15375253
Brand: Techne™ 1610187
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Accurately control liquid bath temperature
Immersion Circulators accurately heat, circulate, and safely control temperature of liquid in the bath within precise limits. Units feature precise PID temperature control, a bright digital LED display, overtemperature indicator light, pressure pump, and low-liquid level cutoff. Unique clamp-on feature allows all circulators to be securely fastened to recommended list of unheated and liquid calibration baths. The TU models offer higher temperatures, increased stability, and an RS-232 interface with TechneWorks software package.
Optional flow coolers used in conjunction with immersion circulators, may be purchased to help cool liquid temperatures down to -35°C. These compact refrigeration units continually extract heat from the bath fluid as it flows through the cooler by means of a builtin heat exchanger.
Specifications
Temperature Range -40°C to +250°C
Type Thermoregulator
Electrical Requirements 120 V
Voltage 120 V