missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUB Recombinant Rabbit Monoclonal Antibody (JE58-75)

TUB Recombinant Rabbit Monoclonal Antibody (JE58-75)
Brand: Invitrogen MA544885
This item is not returnable.
View return policy
Description
TUB Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
| TUB | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P50586, P50607 | |
| TUB | |
| Synthetic peptide within human TUB aa 1-50/506. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| JE58-75 | |
| Unconjugated | |
| TUB | |
| CELE_F10B5.4; F10B5.4; Rd5; RDOB; retinal degeneration 5; TUB; TUB bipartite transcription factor; TUB-1; tubby bipartite transcription factor; tubby candidate gene; tubby homolog; tubby homologue; tubby protein; Tubby protein homolog; Tubby protein homolog 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 22141, 7275 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction